Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-92
Entrez Gene ID
186557
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F59A1.3
Chromosome Location
Chr.V: 17674946 - 17676653 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075072.1 NP_507473.1 2 - 3 491 97

View Full Product Details