Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-31
Entrez Gene ID
189249
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_W06D12.7
Chromosome Location
Chr.V: 17707184 - 17709823 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075078.2 NP_507479.1 2 - 3 641 101

View Full Product Details