Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fat-5
Entrez Gene ID
180162
Gene Name
Delta(9)-fatty-acid desaturase fat-5
Gene Aliases
CELE_W06D12.3
Chromosome Location
Chr.V: 17725004 - 17726818 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075081.5 NP_507482.1 2 - 3 549 64
GenBank mRNA AF260242.1 - 2 - 3 548 64

View Full Product Details