Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-10
Entrez Gene ID
183794
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_C54E10.4
Chromosome Location
Chr.V: 18802995 - 18806509 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075273.4 NP_507674.2 4 - 5 624 67

View Full Product Details