Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp-33D3
Entrez Gene ID
189444
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_Y17D7A.4
Chromosome Location
Chr.V: 18832264 - 18837225 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075278.4 NP_507679.2 4 - 5 895 150

View Full Product Details