Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
emb-4
Entrez Gene ID
180232
Gene Name
protein-coding
Gene Aliases
CELE_Y80D3A.2
Chromosome Location
Chr.V: 18860285 - 18894974 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269902.2 NP_001256831.1 17 - 18 4086 117
NM_001269903.3 NP_001256832.1 2 - 3 435 117
NM_001277003.3 NP_001263932.1 8 - 9 1629 117

View Full Product Details