Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nlp-42
Entrez Gene ID
180235
Gene Name
Neuropeptide-Like Protein
Gene Aliases
CELE_Y80D3A.10
Chromosome Location
Chr.V: 18903772 - 18906252 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075286.5 NP_507687.1 1 - 2 88 70

View Full Product Details