Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-85
Entrez Gene ID
182903
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C25F9.1
Chromosome Location
Chr.V: 19429387 - 19431254 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374928.2 NP_001361855.1 3 - 4 527 92

View Full Product Details