Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-50
Entrez Gene ID
180299
Gene Name
C-type LECtin
Gene Aliases
CELE_W04E12.8
Chromosome Location
Chr.V: 19752399 - 19755368 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075429.8 NP_507830.1 3 - 4 722 78

View Full Product Details