Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-51
Entrez Gene ID
180311
Gene Name
Serine/threonine-protein kinase unc-51
Gene Aliases
CELE_Y60A3A.1
Chromosome Location
Chr.V: 19995942 - 20004485 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075468.6 NP_507869.1 4 - 5 1178 65
GenBank mRNA Z38016.1 - - 1200 65

View Full Product Details