The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029041.3 | NP_001024212.1 | 3 - 4 | 205 | 66 |
NM_001136442.4 | NP_001129914.1 | 2 - 3 | 92 | 66 | |
NM_001330887.2 | NP_001317838.1 | 2 - 3 | 167 | 66 |