Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ttx-1
Entrez Gene ID
180313
Gene Name
protein-coding
Gene Aliases
CELE_Y113G7A.6
Chromosome Location
Chr.V: 20053719 - 20064176 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029042.5 NP_001024213.1 3 - 4 197 68

View Full Product Details