Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fre-1
Entrez Gene ID
180314
Gene Name
protein-coding
Gene Aliases
CELE_Y113G7A.8
Chromosome Location
Chr.V: 20075916 - 20087735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075474.6 NP_507875.1 5 - 6 898 127
GenBank mRNA AY079165.1 - 5 - 6 888 127

View Full Product Details