Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grd-2
Entrez Gene ID
180349
Gene Name
GRounDhog (hedgehog-like family)
Gene Aliases
CELE_F46B3.5
Chromosome Location
Chr.V: 20609794 - 20613929 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075576.4 NP_507977.2 10 - 11 2807 108

View Full Product Details