Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srxa-14
Entrez Gene ID
189894
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y44A6B.1
Chromosome Location
Chr.V: 20639594 - 20641049 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075587.2 NP_507988.2 4 - 5 619 88

View Full Product Details