Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnj-25
Entrez Gene ID
180356
Gene Name
protein-coding
Gene Aliases
CELE_W07A8.3
Chromosome Location
Chr.V: 20729948 - 20744346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270018.2 NP_001256947.1 16 - 17 2231 90
NM_001270019.3 NP_001256948.1 9 - 10 1233 90
NM_001270020.4 NP_001256949.1 8 - 9 1183 90
NM_001373209.3 NP_001360578.1 16 - 17 2244 90
GenBank mRNA AF293972.1 - 16 - 17 2251 90

View Full Product Details