Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srxa-1
Entrez Gene ID
189271
Gene Name
Serpentine Receptor, class XA
Gene Aliases
CELE_W07A8.1
Chromosome Location
Chr.V: 20732087 - 20733241 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075602.2 NP_508003.1 2 - 3 414 132

View Full Product Details