Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdf-9
Entrez Gene ID
189901
Gene Name
Protein sdf-9
Gene Aliases
CELE_Y44A6D.4
Chromosome Location
Chr.V: 20798218 - 20801207 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075612.4 NP_508013.2 4 - 5 622 104
GenBank mRNA AB108783.1 - 4 - 5 644 104

View Full Product Details