Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-22
Entrez Gene ID
180365
Gene Name
protein-coding
Gene Aliases
CELE_T03D8.5
Chromosome Location
Chr.V: 20820118 - 20825062 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277022.3 NP_001263951.1 16 - 17 3073 99
NM_001277023.4 NP_001263952.1 16 - 17 3085 99
NM_075617.5 NP_508018.2 16 - 17 3131 99

View Full Product Details