Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
num-1
Entrez Gene ID
180367
Gene Name
protein-coding
Gene Aliases
CELE_T03D8.1
Chromosome Location
Chr.V: 20829254 - 20837778 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028926.5 NP_001024097.1 6 - 7 2656 80
NM_001028927.3 NP_001024098.1 7 - 8 1519 80
NM_001136432.5 NP_001129904.1 9 - 10 2475 80
NM_001270029.3 NP_001256958.1 6 - 7 1477 80
NM_075620.6 NP_508021.1 7 - 8 1651 80
GenBank mRNA AF286205.1 - 7 - 8 1651 80
AF286206.1 - 7 - 8 1519 80

View Full Product Details