Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-35
Entrez Gene ID
185149
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F31D4.7
Chromosome Location
Chr.V: 20862981 - 20865795 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075630.6 NP_508031.3 7 - 8 1259 86

View Full Product Details