Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-28
Entrez Gene ID
190641
Gene Name
SET domain-containing protein
Gene Aliases
CELE_Y73B3B.2
Chromosome Location
Chr.X: 214580 - 215588 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075648.2 NP_508049.1 1 - 2 574 81

View Full Product Details