Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nrde-3
Entrez Gene ID
180392
Gene Name
Nuclear RNAi defective-3 protein
Gene Aliases
CELE_R04A9.2
Chromosome Location
Chr.X: 372235 - 376900 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075691.7 NP_508092.1 1 - 2 282 89

View Full Product Details