Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dve-1
Entrez Gene ID
180398
Gene Name
protein-coding
Gene Aliases
CELE_ZK1193.5
Chromosome Location
Chr.X: 427035 - 437236 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029813.6 NP_001024984.1 2 - 3 294 76
NM_001351875.4 NP_001338861.1 1 - 1 180 76
NR_147826.1 1 - 2 180 76

View Full Product Details