Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dve-1
Entrez Gene ID
180398
Gene Name
protein-coding
Gene Aliases
CELE_ZK1193.5
Chromosome Location
Chr.X: 427035 - 437236 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029813.6 NP_001024984.1 9 - 10 1435 95
NM_001351875.4 NP_001338861.1 8 - 9 1312 95
NM_001351876.3 NP_001338860.1 8 - 9 1320 95
NM_001383518.1 9 - 10 1436 95
NR_147826.1 7 - 8 2015 95

View Full Product Details