Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ugt-46
Entrez Gene ID
180404
Gene Name
protein-coding
Gene Aliases
CELE_B0310.5
Chromosome Location
Chr.X: 525928 - 530289 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075711.7 NP_508112.1 3 - 4 650 80

View Full Product Details