Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irk-2
Entrez Gene ID
191695
Gene Name
protein-coding
Gene Aliases
CELE_M02A10.2
Chromosome Location
Chr.X: 702971 - 714498 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270047.4 NP_001256976.2 3 - 4 294 124
NM_001270048.3 NP_001256977.1 3 - 4 302 124
NM_001307921.4 NP_001294850.1 3 - 4 304 124
NM_001330888.3 NP_001317907.1 4 - 5 393 124
NM_001380895.1 NP_001368610.1 4 - 5 393 124
NM_001380896.1 NP_001368348.1 4 - 5 413 124

View Full Product Details