Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifd-2
Entrez Gene ID
180430
Gene Name
Intermediate filament protein ifd-2
Gene Aliases
CELE_F25E2.4
Chromosome Location
Chr.X: 813851 - 815929 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075759.6 NP_508160.1 4 - 5 521 101

View Full Product Details