Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-3
Entrez Gene ID
180431
Gene Name
protein-coding
Gene Aliases
CELE_F25E2.5
Chromosome Location
Chr.X: 817922 - 825475 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029434.5 NP_001024605.1 4 - 5 703 92
NM_001313414.3 NP_001300343.1 6 - 7 933 92
NM_001380898.1 NP_001368578.1 4 - 5 650 92
NM_075760.6 NP_508161.3 7 - 8 1035 92
GenBank mRNA AF005205.1 - 4 - 5 698 92

View Full Product Details