Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
igcm-1
Entrez Gene ID
180433
Gene Name
ImmunoGlobulin-like Cell adhesion Molecule family
Gene Aliases
CELE_F39H12.4
Chromosome Location
Chr.X: 845731 - 851525 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075765.7 NP_508166.1 6 - 7 1043 114

View Full Product Details