Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-11
Entrez Gene ID
180444
Gene Name
protein-coding
Gene Aliases
CELE_F35H12.2
Chromosome Location
Chr.X: 924958 - 932261 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373217.4 NP_001360743.1 4 - 5 363 146
NM_075777.7 NP_508178.2 4 - 5 317 146
NM_075778.5 NP_508179.2 4 - 5 370 146

View Full Product Details