Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apm-3
Entrez Gene ID
186194
Gene Name
MHD domain-containing protein
Gene Aliases
CELE_F53H8.1
Chromosome Location
Chr.X: 954839 - 957762 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075783.6 NP_508184.2 7 - 8 965 105

View Full Product Details