Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncs-3
Entrez Gene ID
186953
Gene Name
Neuronal Calcium Sensor family
Gene Aliases
CELE_K03E6.3
Chromosome Location
Chr.X: 1054081 - 1056849 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075800.5 NP_508201.2 3 - 4 474 63

View Full Product Details