Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lim-6
Entrez Gene ID
180459
Gene Name
protein-coding
Gene Aliases
CELE_K03E6.1
Chromosome Location
Chr.X: 1073908 - 1079452 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270051.2 NP_001256980.1 4 - 5 599 83
NM_001270052.2 NP_001256981.1 4 - 5 357 83

View Full Product Details