Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cebp-1
Entrez Gene ID
180481
Gene Name
C/EBP (CCAAT/enhancer-binding protein) homolog
Gene Aliases
CELE_D1005.3
Chromosome Location
Chr.X: 1452202 - 1453868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075875.6 NP_508276.2 1 - 2 158 53

View Full Product Details