Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wrt-9
Entrez Gene ID
181931
Gene Name
WaRThog (hedgehog-like family)
Gene Aliases
CELE_B0344.2
Chromosome Location
Chr.X: 1576637 - 1591321 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075897.5 NP_508298.2 10 - 11 1475 59

View Full Product Details