Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dgn-3
Entrez Gene ID
184149
Gene Name
Peptidase S72 domain-containing protein
Gene Aliases
CELE_F07G6.1
Chromosome Location
Chr.X: 1729858 - 1733447 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075918.3 NP_508319.3 1 - 2 314 142

View Full Product Details