The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001383530.1 | 2 - 3 | 470 | 71 | |
GenBank mRNA | AJ427855.1 | - | 2 - 3 | 472 | 71 |