Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx-41
Entrez Gene ID
188917
Gene Name
Putative T-box protein 41
Gene Aliases
CELE_T26C11.1
Chromosome Location
Chr.X: 1865246 - 1866638 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_138513.1 2 - 3 290 94

View Full Product Details