Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lsy-2
Entrez Gene ID
180522
Gene Name
protein-coding
Gene Aliases
CELE_F49H12.1
Chromosome Location
Chr.X: 2089659 - 2093256 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029525.4 NP_001024696.1 2 - 3 307 78
NM_001029526.4 NP_001024697.1 1 - 2 139 78

View Full Product Details