Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
peb-1
Entrez Gene ID
180532
Gene Name
protein-coding
Gene Aliases
CELE_T14F9.4
Chromosome Location
Chr.X: 2211051 - 2218988 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029739.6 NP_001024910.1 1 - 2 128 152
NM_001377612.2 NP_001364564.1 2 - 3 173 152

View Full Product Details