Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-15
Entrez Gene ID
180534
Gene Name
Probable V-type proton ATPase subunit H 2
Gene Aliases
CELE_T14F9.1
Chromosome Location
Chr.X: 2237911 - 2240255 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076011.4 NP_508412.1 1 - 2 78 66

View Full Product Details