Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glr-7
Entrez Gene ID
180547
Gene Name
protein-coding
Gene Aliases
CELE_C43H6.9
Chromosome Location
Chr.X: 2413909 - 2419877 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001350348.3 NP_001337310.1 11 - 12 1804 102
NM_001373243.1 NP_001359931.1 7 - 8 1255 102

View Full Product Details