Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-31
Entrez Gene ID
191655
Gene Name
protein-coding
Gene Aliases
CELE_T07D1.1
Chromosome Location
Chr.X: 2419955 - 2427504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029717.2 NP_001024888.1 8 - 9 1116 128
NM_001029718.2 NP_001024889.1 11 - 12 1440 128
NM_001029719.1 NP_001024890.1 9 - 10 1068 128
NM_001330893.1 NP_001317860.1 10 - 11 1329 128
GenBank mRNA AY275182.1 - 11 - 12 1440 128
AY275183.1 - 9 - 10 1068 128
AY275184.1 - 8 - 9 1116 128

View Full Product Details