Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
R04B3.2
Entrez Gene ID
180550
Gene Name
Glycosylasparaginase beta chain
Gene Aliases
CELE_R04B3.2
Chromosome Location
Chr.X: 2471540 - 2473718 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076047.5 NP_508448.1 5 - 6 843 65

View Full Product Details