Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbc-12
Entrez Gene ID
180556
Gene Name
Rab-GAP TBC domain-containing protein
Gene Aliases
CELE_R11B5.1
Chromosome Location
Chr.X: 2525191 - 2532457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076057.4 NP_508458.1 2 - 3 342 80

View Full Product Details