Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ssr-2
Entrez Gene ID
191965
Gene Name
Sperm-Specific family, class R
Gene Aliases
CELE_C14A11.7
Chromosome Location
Chr.X: 2823501 - 2827752 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076089.6 NP_508490.4 2 - 3 192 88

View Full Product Details