Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tra-4
Entrez Gene ID
180575
Gene Name
TRAnsformer: XX animals transformed into males
Gene Aliases
CELE_F53B3.1
Chromosome Location
Chr.X: 2853674 - 2857566 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383535.1 6 - 7 1182 69

View Full Product Details