Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmy-1
Entrez Gene ID
180581
Gene Name
protein-coding
Gene Aliases
CELE_F52B10.1
Chromosome Location
Chr.X: 2909527 - 2918044 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374981.3 NP_001362097.1 3 - 4 510 66
NM_076103.6 NP_508504.2 3 - 4 511 66

View Full Product Details