Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osm-5
Entrez Gene ID
180585
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y41G9A.1
Chromosome Location
Chr.X: 2984263 - 2991478 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076110.7 NP_508511.1 12 - 13 1934 129
GenBank mRNA AF314195.1 - 12 - 13 1919 129

View Full Product Details